Learn More
Microsolv Technology Corporation Vials 10Ml 46 X 22.5 Crm Ctc A
Catalog No. 5001451887
Encompass_Preferred
Vials, Headspace, Crimp Top, Glass. Amber, 10ml, 20mm Serum Finish, 22.5x46mm outer dimensions, Rounded Bottom. CTC Compliant. MicroSolv Brand. 100/PK.
Catalog No. 50-014-51887 Supplier Microsolv Technology Corporation Supplier No. 95025CT10A
Vials, Headspace, Crimp Top, Glass. Amber, 10ml, 20mm Serum Finish, 22.5x46mm outer dimensions, Rounded Bottom. CTC Compliant. MicroSolv Brand. 100/PK.