Learn More
Microsolv Technology Corporation Vials 10Ml 46 X 22.5Mm Crp Agc
Catalog No. 5001451881
Encompass_Preferred
Vials, Headspace, Crimp Top, Glass. Clear, 10ml, 20mm Serum Finish, 22.5x46mm outer dimensions, Flat Bottom with a Long Neck. Agilent Compatible. MicroSolv Brand. 100/PK.
Catalog No. 50-014-51881 Supplier Microsolv Technology Corporation Supplier No. 95025AG10C
Vials, Headspace, Crimp Top, Glass. Clear, 10ml, 20mm Serum Finish, 22.5x46mm outer dimensions, Flat Bottom with a Long Neck. Agilent Compatible. MicroSolv Brand. 100/PK.