Learn More
Microsolv Technology Corporation Vials 20Ml 75.5 X 23Mm Crimpwo
Catalog No. 5001451900
Encompass_Preferred
Vials, Headspace, Crimp Top, Glass. Clear with Write-On Patch and fill lines. 20ml, 20mm Serum Finish, 23x75.5mm outer dimensions, Beveled Top and Rounded Bottom with a Tapered Neck. Perkin Elmer Compatible. MicroSolv Brand. 100/PK.
Catalog No. 50-014-51900 Supplier Microsolv Technology Corporation Supplier No. 95025PE20WC
Vials, Headspace, Crimp Top, Glass. Clear with Write-On Patch and fill lines. 20ml, 20mm Serum Finish, 23x75.5mm outer dimensions, Beveled Top and Rounded Bottom with a Tapered Neck. Perkin Elmer Compatible. MicroSolv Brand. 100/PK.