Learn More
Microsolv Technology Corporation Vials 20Ml 75.5X 22.5 Cr Ctc C
Catalog No. 5001451890
Encompass_Preferred
Vials, Headspace, Crimp Top, Glass. Clear, 20ml, 20mm Serum Finish, 22.5x75.5mm outer dimensions, Rounded Bottom. CTC Compliant. MicroSolv Brand. 100/PK.
Catalog No. 50-014-51890 Supplier Microsolv Technology Corporation Supplier No. 95025CT20C
Vials, Headspace, Crimp Top, Glass. Clear, 20ml, 20mm Serum Finish, 22.5x75.5mm outer dimensions, Rounded Bottom. CTC Compliant. MicroSolv Brand. 100/PK.