Learn More
Microsolv Technology Corporation Vials, Headspace, Crimp Top, Glass. Clear. 20ml, 20mm Serum Finish, 23x75mm outer dimensions, Beveled Top and Rounded Bottom with a Tapered Neck. Perkin Elmer Compatible. 100/PK.
Catalog No. 5001451896
Encompass_Preferred
First hydrolytic, Class A, Borosilicate Glass. Use with aluminum or steel crimp caps for proper sealing. Compatible with Perkin Elmer, Tekmar and other Headspace autosamplers.
Catalog No. 50-014-51896 Supplier Microsolv Technology Corporation Supplier No. 95025EPE20C
First hydrolytic, Class A, Borosilicate Glass. Use with aluminum or steel crimp caps for proper sealing. Compatible with Perkin Elmer, Tekmar and other Headspace autosamplers.