Learn More
Thomas Scientific Scintillation Vials - 20ml, 20ml vials and caps, 5 trays of 100
SDP
Catalog No. 501461745
Encompass
Leakproof, 1¼ turn cap Wide mouth - 24mm opening Lower background than glass These standard size vials fit all major counters. The linerless polythylene cap goes on the polypropylene vial easily, yet seals completely. Less than 50% the background of glass. Suitable for low activity counting. Caps and vials packed separately, 500 vials per case
Catalog No. 50-146-1745 Supplier Thomas Scientific Supplier No. 1159N23
Leakproof, 1¼ turn cap Wide mouth - 24mm opening Lower background than glass These standard size vials fit all major counters. The linerless polythylene cap goes on the polypropylene vial easily, yet seals completely. Less than 50% the background of glass. Suitable for low activity counting. Caps and vials packed separately, 500 vials per case