Learn More
Microsolv Technology Corporation Vials-300Ul Clear 12X32Mm
Catalog No. NC1544060
Encompass_Preferred
Vials, Screw Top, Glass. Clear / Clear Insert, 300ul with Write-On Patch and fill lines, Wide Tip Fused Insert™, 9-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial. RSA Brand. 100/PK.
Catalog No. NC1544060 Supplier Microsolv Technology Corporation Supplier No. 9532SWCVWTRS
Vials, Screw Top, Glass. Clear / Clear Insert, 300ul with Write-On Patch and fill lines, Wide Tip Fused Insert™, 9-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial. RSA Brand. 100/PK.