Learn More
Microsolv Technology Corporation Vials 40Ml Epa Clear 100/Pk
Catalog No. 5001452439
Encompass_Preferred
Vials, EPA Style, Glass. Clear, 40ml. 95.5x27.5mm outer dimensions. For use as an EPA test vial or storage bottle. ND24 threads. MicroSolv Brand. 100/PK.
Catalog No. 50-014-52439 Supplier Microsolv Technology Corporation Supplier No. 970400CV
Vials, EPA Style, Glass. Clear, 40ml. 95.5x27.5mm outer dimensions. For use as an EPA test vial or storage bottle. ND24 threads. MicroSolv Brand. 100/PK.