Learn More
Microsolv Technology Corporation Vials 5Ml 38.2 X 22Mm Crimp
Catalog No. 5001451897
Encompass_Preferred
Vials, Headspace, Crimp Top, Glass. Clear, 5ml, 20mm Serum Finish, 22x38.2mm outer dimensions, Rounded Bottom. Perkin Elmer Compliant. MicroSolv Brand. 100/PK.
Catalog No. 50-014-51897 Supplier Microsolv Technology Corporation Supplier No. 95025PE05C
Vials, Headspace, Crimp Top, Glass. Clear, 5ml, 20mm Serum Finish, 22x38.2mm outer dimensions, Rounded Bottom. Perkin Elmer Compliant. MicroSolv Brand. 100/PK.