Learn More
Microsolv Technology Corporation Vials 5Ml 38 X 22Mm Crm V C
Catalog No. 5001451903
Encompass_Preferred
Vials, Headspace, Crimp Top, Glass. Clear. 5ml, 20mm Serum Finish, 20x38mm outer dimensions, Flat Bottom. Varian Compliant. MicroSolv Brand. 100/PK.
Catalog No. 50-014-51903 Supplier Microsolv Technology Corporation Supplier No. 95025VN05C
Vials, Headspace, Crimp Top, Glass. Clear. 5ml, 20mm Serum Finish, 20x38mm outer dimensions, Flat Bottom. Varian Compliant. MicroSolv Brand. 100/PK.