Learn More
Chemglass Life Sciences Chemglass Autosampler Vials, Clear Glass, 6.0 mL, GPI 18-400, 100/Pack
SDP
Catalog No. NC1920276
Encompass_Preferred
Chemglass 6.0 mL clear glass autosampler vials are designed for automated sampling systems and laboratory analysis. They provide reliable sample containment and are supplied in convenient 100-vial packs for routine autosampler workflows.
  • 6.0 mL capacity.
  • Borosilicate glass construction.
  • Clear for optical clarity.
  • GPI 18-400 thread for standard closures.
  • Pack of 100 vials per pack.
  • Designed for compatibility with autosamplers and automated systems.
Catalog No. NC1920276 Supplier Chemglass Life Sciences Supplier No. CV11502347
Chemglass 6.0 mL clear glass autosampler vials are designed for automated sampling systems and laboratory analysis. They provide reliable sample containment and are supplied in convenient 100-vial packs for routine autosampler workflows.
  • 6.0 mL capacity.
  • Borosilicate glass construction.
  • Clear for optical clarity.
  • GPI 18-400 thread for standard closures.
  • Pack of 100 vials per pack.
  • Designed for compatibility with autosamplers and automated systems.