Learn More
Microsolv Technology Corporation Vials 60Ml Epa Amber 100/Pk
Catalog No. 5001452445
Encompass_Preferred
Vials, EPA Style, Glass. Amber, 60ml. 140x27.5mm outer dimensions. For use as an EPA test vial or storage bottle. ND24 threads. MicroSolv Brand. 100/PK.
Catalog No. 50-014-52445 Supplier Microsolv Technology Corporation Supplier No. 970600AV
Vials, EPA Style, Glass. Amber, 60ml. 140x27.5mm outer dimensions. For use as an EPA test vial or storage bottle. ND24 threads. MicroSolv Brand. 100/PK.