Learn More
Microsolv Technology Corporation Vials, Headspace, Crimp Top, Glass. Clear, 6ml, 20mm Serum Finish, 22x38mm outer dimensions, Beveled Top and Flat Bottom with a Tapered Neck. Agilent Compatible. 100/PK.
Catalog No. 5001451883
Encompass_Preferred
First hydrolytic, Class A, Borosilicate Glass, Use with aluminum or steel crimp caps for proper sealing.
Catalog No. 50-014-51883 Supplier Microsolv Technology Corporation Supplier No. 95025AP06C
First hydrolytic, Class A, Borosilicate Glass, Use with aluminum or steel crimp caps for proper sealing.