Learn More
Microsolv Technology Corporation Vials Amber With Write On
Catalog No. 5001451920
Encompass_Preferred
Vials, Crimp Top, Glass. Amber, 2ml, with Write-On Patch and fill lines. An 11mm Serum Finish with 12x32mm outer dimensions. For use as an autosampler vial. MicroSolv Brand. 100/PK.
Catalog No. 50-014-51920 Supplier Microsolv Technology Corporation Supplier No. 9502C0WAV
Vials, Crimp Top, Glass. Amber, 2ml, with Write-On Patch and fill lines. An 11mm Serum Finish with 12x32mm outer dimensions. For use as an autosampler vial. MicroSolv Brand. 100/PK.