Learn More
Microsolv Technology Corporation Vials And Caps Shell 1Ml Clear
Catalog No. 5001451815
Encompass_Preferred
Vial & Cap Kit includes 100 EA 1ml, Push Top, Clear Autosampler Vials and 100 EA matching, translucent, polyethylene Push Caps for Micro-Insert. MicroSolv Brand Easy Purchase Pack. 1/PK.
Catalog No. 50-014-51815 Supplier Microsolv Technology Corporation Supplier No. 95001CSVEPP
Vial & Cap Kit includes 100 EA 1ml, Push Top, Clear Autosampler Vials and 100 EA matching, translucent, polyethylene Push Caps for Micro-Insert. MicroSolv Brand Easy Purchase Pack. 1/PK.