Learn More
Chemglass Life Sciences Vial, 1.2mL, MRQ30, High Recovery, 9mm Thread, w/ PTFE/Silicone Bonded Cap
SDP
Catalog No. 50230620
Encompass_Preferred
Vial, 1.2mL, MRQ30, High Recovery, 9mm Thread, w/ PTFE/Silicone Bonded Cap. This patent pending design incorporates a unique, continuous wall that mimics a mandrel point insert at the bottom of the vial for extreme reproducibility. These vials are useful when you want to inject almost al contents of the vials. Excellent choice for standards. Available ¿deactivated¿ for compounds that stick to or react with glass. Sold as a convenience pack with a 9mm, PTFE/Silicone bonded screw cap.
Catalog No. 50-230-620 Supplier Chemglass Life Sciences Supplier No. CV51921232
Vial, 1.2mL, MRQ30, High Recovery, 9mm Thread, w/ PTFE/Silicone Bonded Cap. This patent pending design incorporates a unique, continuous wall that mimics a mandrel point insert at the bottom of the vial for extreme reproducibility. These vials are useful when you want to inject almost al contents of the vials. Excellent choice for standards. Available ¿deactivated¿ for compounds that stick to or react with glass. Sold as a convenience pack with a 9mm, PTFE/Silicone bonded screw cap.