Learn More
Microsolv Technology Corporation Vials Clear 1.5Ml 12X32Mm
Catalog No. 5001451906
Encompass_Preferred
Vials, Crimp Top, Glass. Clear, 2ml. An 11mm Serum Finish with 12x32mm outer dimensions. For use as an autosampler vial or storage bottle. MicroSolv Brand. 100/PK.
Catalog No. 50-014-51906 Supplier Microsolv Technology Corporation Supplier No. 9502C00CV
Vials, Crimp Top, Glass. Clear, 2ml. An 11mm Serum Finish with 12x32mm outer dimensions. For use as an autosampler vial or storage bottle. MicroSolv Brand. 100/PK.