Learn More
Chemglass Life Sciences Pack, Blue Bonded Closure and Vial, Glass, 2.0mL, 9mm Thread, Clear, 12 x 32mm, With PTFE/Red Rubber Septa, 3 Packs of 100
SDP
Catalog No. 501311405
Encompass_Preferred
Pack, Blue Bonded Closure and Vial, Glass, 2.0mL, 9mm Thread, Clear, 12 x 32mm, With PTFE/Red Rubber Septa, 3 Packs of 100.• Convenience packs with 2.0mL, 12 x 32mm clear vials• I-D vials have marking spots and graduations (0.5, 1.0 and 1.5mL)• Proper fit assured - caps and vials made to the same specification• Supplied in two-compartment storage trays• Septa fallout common with pre-assembled closures is eliminated• Choose PTFE/red rubber or PTFE/silicone septa• Septa available with or without slitsEasy to Order, Easy to Save!We've made it as easy as possible for you to enjoy significant savings while experiencing the performance of a great little product. If you've never tried vials with bonded closures or if you're already a committed user, just specify the part number from the table below and you'll receive three convenience packs for the two-pack price.
Catalog No. 50-131-1405 Supplier Chemglass Life Sciences Supplier No. CV400080S
Pack, Blue Bonded Closure and Vial, Glass, 2.0mL, 9mm Thread, Clear, 12 x 32mm, With PTFE/Red Rubber Septa, 3 Packs of 100.• Convenience packs with 2.0mL, 12 x 32mm clear vials• I-D vials have marking spots and graduations (0.5, 1.0 and 1.5mL)• Proper fit assured - caps and vials made to the same specification• Supplied in two-compartment storage trays• Septa fallout common with pre-assembled closures is eliminated• Choose PTFE/red rubber or PTFE/silicone septa• Septa available with or without slitsEasy to Order, Easy to Save!We've made it as easy as possible for you to enjoy significant savings while experiencing the performance of a great little product. If you've never tried vials with bonded closures or if you're already a committed user, just specify the part number from the table below and you'll receive three convenience packs for the two-pack price.