Learn More
Microsolv Technology Corporation Vials Clear Crimp Top 1.5Ml
Catalog No. 5001451919
Encompass_Preferred
Vials, Crimp Top, Glass. Clear, 2ml. An 11mm Serum Finish with 12x32mm outer dimensions. For use as an autosampler vial or storage bottle. BASIK Brand. 100/PK.
Catalog No. 50-014-51919 Supplier Microsolv Technology Corporation Supplier No. 9502C0S
Vials, Crimp Top, Glass. Clear, 2ml. An 11mm Serum Finish with 12x32mm outer dimensions. For use as an autosampler vial or storage bottle. BASIK Brand. 100/PK.