Learn More
Microsolv Technology Corporation Vials Clr W-On4Ml 15X45 100/Pk
Catalog No. 5001452240
Encompass_Preferred
Vials, Screw Top, Glass. Clear, 4ml, with Write-On Patch and fill lines, 13-425mm threads, 15x45mm outer dimensions. For use as an autosampler vial or storage bottle. MicroSolv Brand. 100/PK.
Catalog No. 50-014-52240 Supplier Microsolv Technology Corporation Supplier No. 9504SWCV
Vials, Screw Top, Glass. Clear, 4ml, with Write-On Patch and fill lines, 13-425mm threads, 15x45mm outer dimensions. For use as an autosampler vial or storage bottle. MicroSolv Brand. 100/PK.