Learn More
Microsolv Technology Corporation Vials Clr Wo W/Fused Inserts
Catalog No. 5001452370
Encompass_Preferred
Vials, Screw Top, Glass. Clear / Clear Insert, 300ul with Write-On Patch and fill lines, Fused Insert™, 9-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial. AQ Brand. 100/PK.
Catalog No. 50-014-52370 Supplier Microsolv Technology Corporation Supplier No. 9532S0CV
Vials, Screw Top, Glass. Clear / Clear Insert, 300ul with Write-On Patch and fill lines, Fused Insert™, 9-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial. AQ Brand. 100/PK.