Learn More
Microsolv Technology Corporation Vials, Crimp Top, Glass. Clear / Clear Insert, 300ul, with Write-On Patch and fill lines, Fused Insert Style. An 11mm Serum Finish with 12x32mm outer dimensions. For use as an autosampler vial. 100/PK.
Catalog No. 502271363
Encompass_Preferred
Vials and Inserts are made with first hydrolytic, Class A, Type-N51A Clear, Borosilicate Glass. The insert has a 300ul total fill volume and a 3ul residual volume for many autosamplers and autosampler needles. The Write-On Patch and fill lines are epoxy paint and will resist smearing when working with solvents. Use with 11mm aluminum crimp caps. These lower cost Fused Insert vials have serum finish tops that provide the best seal for chromatography, when used properly with correct aluminum crimp caps.
This item is not returnable. View return policy
Catalog No. 50-227-1363 Supplier Microsolv Technology Corporation Supplier No. 9532C0WCV
Vials and Inserts are made with first hydrolytic, Class A, Type-N51A Clear, Borosilicate Glass. The insert has a 300ul total fill volume and a 3ul residual volume for many autosamplers and autosampler needles. The Write-On Patch and fill lines are epoxy paint and will resist smearing when working with solvents. Use with 11mm aluminum crimp caps. These lower cost Fused Insert vials have serum finish tops that provide the best seal for chromatography, when used properly with correct aluminum crimp caps.