Learn More
Microsolv Technology Corporation Vials, Crimp Top, Glass. Amber, 2ml. Deactivated. An 11mm Serum Finish with 12x32mm outer dimensions. For use as an autosampler vial. 100/PK.
Catalog No. 502270972
Encompass_Preferred
First hydrolytic, Class A, Deactivated / Silanized Borosilicate Glass, amberized. Amber vials are often used for light sensitive compounds and considered low actinic. Use with 11mm aluminum crimp caps. These vials have serum finish tops that provide the best seal for chromatography, when used properly with correct aluminum crimp caps.
This item is not returnable. View return policy
Catalog No. 50-227-0972 Supplier Microsolv Technology Corporation Supplier No. 9502C0E0DAV
First hydrolytic, Class A, Deactivated / Silanized Borosilicate Glass, amberized. Amber vials are often used for light sensitive compounds and considered low actinic. Use with 11mm aluminum crimp caps. These vials have serum finish tops that provide the best seal for chromatography, when used properly with correct aluminum crimp caps.