Learn More
Microsolv Technology Corporation Vials, Crimp Top, Glass. Amber, 2ml, with Write-On Patch and fill lines. An 11mm Serum Finish with 12x32mm outer dimensions. For use as an autosampler vial or storage bottle. 100/PK.
Catalog No. 502271608
Encompass_Preferred
First hydrolytic, Class A, Borosilicate Glass, amberized. Amber vials are often used for light sensitive compounds and considered low actinic. Write-On Patch and fill lines are epoxy paint for reduced smearing with solvents. Use with 11mm aluminum crimp caps. These vials have serum finish tops that provide the best seal for chromatography, when used properly with correct aluminum crimp caps.
This item is not returnable. View return policy
Catalog No. 50-227-1608 Supplier Microsolv Technology Corporation Supplier No. 9502C0EWAV
First hydrolytic, Class A, Borosilicate Glass, amberized. Amber vials are often used for light sensitive compounds and considered low actinic. Write-On Patch and fill lines are epoxy paint for reduced smearing with solvents. Use with 11mm aluminum crimp caps. These vials have serum finish tops that provide the best seal for chromatography, when used properly with correct aluminum crimp caps.