Learn More
Microsolv Technology Corporation Vials, Crimp Top, Glass. Clear, 2ml. An 11mm Serum Finish with 12x32mm outer dimensions. For use as an autosampler vial or storage bottle. 100/PK.
Catalog No. 502270971
Encompass_Preferred
First hydrolytic, Class A, Borosilicate Glass. Use with 11mm aluminum crimp caps. These vials have serum finish tops that provide the best seal for chromatography, when used properly with correct aluminum crimp caps.
This item is not returnable. View return policy
Catalog No. 50-227-0971 Supplier Microsolv Technology Corporation Supplier No. 9502C0E0CV
First hydrolytic, Class A, Borosilicate Glass. Use with 11mm aluminum crimp caps. These vials have serum finish tops that provide the best seal for chromatography, when used properly with correct aluminum crimp caps.