Learn More
Microsolv Technology Corporation Vials, Crimp Top, Glass. Clear, 1.5ml, U Shape. An 11mm Serum Finish with 12x32mm outer dimensions and a fluted stand. For use as an autosampler vial. 100/PK.
Catalog No. 502271397
Encompass_Preferred
First hydrolytic, Class A, Type N51A, Borosilicate Glass. Compares to Champagne Style Vials. Features a 15ul residual volume for most autosamplers due to the center draining, inner U shape. Use with 11mm aluminum crimp caps. These lower cost center draining vials have serum finish tops that provide the best seal for chromatography, when used properly with correct aluminum crimp caps.
This item is not returnable. View return policy
Catalog No. 50-227-1397 Supplier Microsolv Technology Corporation Supplier No. 9512C00CVP
First hydrolytic, Class A, Type N51A, Borosilicate Glass. Compares to Champagne Style Vials. Features a 15ul residual volume for most autosamplers due to the center draining, inner U shape. Use with 11mm aluminum crimp caps. These lower cost center draining vials have serum finish tops that provide the best seal for chromatography, when used properly with correct aluminum crimp caps.