Learn More
Microsolv Technology Corporation Vials, Crimp Top, Glass. Clear, 1.8ml, Max Recovery Style. An 11mm Serum Finish with 12x32mm outer dimensions and rounded bottom. For use as an autosampler vial. 100/PK.
Catalog No. 502271398
Encompass_Preferred
First hydrolytic, Class A, Type N51A, Borosilicate Glass. Features an 25ul residual volume for most autosamplers due to the center draining interior and conical bottom. Rounded exterior bottom helps when inserting vials into racks or autosamplers with tight fittings. Use with 11mm aluminum crimp caps. These lower cost, center draining vials have serum finish tops that provide the best seal for chromatography, when used properly with correct aluminum crimp caps. Lowest cost in class for budget minded laboratories.
This item is not returnable. View return policy
Catalog No. 50-227-1398 Supplier Microsolv Technology Corporation Supplier No. 9512C0E0CVH
First hydrolytic, Class A, Type N51A, Borosilicate Glass. Features an 25ul residual volume for most autosamplers due to the center draining interior and conical bottom. Rounded exterior bottom helps when inserting vials into racks or autosamplers with tight fittings. Use with 11mm aluminum crimp caps. These lower cost, center draining vials have serum finish tops that provide the best seal for chromatography, when used properly with correct aluminum crimp caps. Lowest cost in class for budget minded laboratories.