Learn More
Microsolv Technology Corporation Vials, Crimp Top, Glass. Clear, 2ml, with Write-On Patch and fill lines. An 11mm Serum Finish with 12x32mm outer dimensions. For use as an autosampler vial or storage bottle. 100/PK.
Catalog No. 502271225
Encompass_Preferred
First hydrolytic, Class A, Type N51A, pharmaceutical grade, Borosilicate Glass. These wide mouth, vials have a Write-On Patch and fill lines that are epoxy paint for reduced smearing with solvents. This is our Crimp Top Vial with a serum finish.
This item is not returnable. View return policy
Catalog No. 50-227-1225 Supplier Microsolv Technology Corporation Supplier No. 9502C0WCV
First hydrolytic, Class A, Type N51A, pharmaceutical grade, Borosilicate Glass. These wide mouth, vials have a Write-On Patch and fill lines that are epoxy paint for reduced smearing with solvents. This is our Crimp Top Vial with a serum finish.