Learn More
Microsolv Technology Corporation Vials, Crimp Top, Glass. Clear, 0.9ml, Tear Drop shape MicroVial, 11mm, 10x32mm outer dimensions. For use as an autosampler vial. 100/PK.
Catalog No. 502271222
Encompass_Preferred
Vials, Crimp Top, Glass. Clear, 0.9ml, Tear Drop shape MicroVial, 11mm, 10x32mm outer dimensions. For use as an autosampler vial. 100/PK.
This item is not returnable. View return policy
Catalog No. 50-227-1222 Supplier Microsolv Technology Corporation Supplier No. 9502C00.9
Vials, Crimp Top, Glass. Clear, 0.9ml, Tear Drop shape MicroVial, 11mm, 10x32mm outer dimensions. For use as an autosampler vial. 100/PK.