Learn More
Microsolv Technology Corporation Vials, Crimp Top, Glass. Clear, 0.2ml. An 8mm Serum Finish with 5.5x31.5mm outer dimensions and a conical bottom. Deactivated. For use as an autosampler vial. 1000/CS.
Catalog No. 502270514
Encompass_Preferred
First hydrolytic, Class A, pharmaceutical grade, Deactivated / Silanized Borosilicate Glass. Use with 8mm aluminum crimp caps, 8mm or 9mm push caps. These conical vials have serum finish tops that provide the best seal for chromatography, when used properly with correct aluminum crimp caps. These MicroVials have a conical outer shape and often need a holder / adapter to be used in an autosampler and will not stand by themselves.
This item is not returnable. View return policy
Catalog No. 50-227-0514 Supplier Microsolv Technology Corporation Supplier No. 9508C0DCV
First hydrolytic, Class A, pharmaceutical grade, Deactivated / Silanized Borosilicate Glass. Use with 8mm aluminum crimp caps, 8mm or 9mm push caps. These conical vials have serum finish tops that provide the best seal for chromatography, when used properly with correct aluminum crimp caps. These MicroVials have a conical outer shape and often need a holder / adapter to be used in an autosampler and will not stand by themselves.