Learn More
Microsolv Technology Corporation Vials, Crimp Top, Glass. Clear, 1.8ml, Max Recovery™. Deactivated. An 11mm Serum Finish with 12x32mm outer dimensions. For use as an autosampler vial. 100/PK.
Catalog No. 502270569
Encompass_Preferred
First hydrolytic, Class A, Type 33, Deactivated / Silanized Borosilicate Glass. Features an 8ul residual volume for most autosamplers due to the center draining interior, smooth walls (no ledges) and the conical bottom. This product is manufactured in an ISO 9001:2015 Certified facility. Compares to Agilent Maximum Recovery vials. Use with 11mm aluminum crimp caps. These lower cost center draining vials have serum finish tops that provide the best seal for chromatography, when used properly with correct aluminum crimp caps.
This item is not returnable. View return policy
Catalog No. 50-227-0569 Supplier Microsolv Technology Corporation Supplier No. 9512C00DCVA
First hydrolytic, Class A, Type 33, Deactivated / Silanized Borosilicate Glass. Features an 8ul residual volume for most autosamplers due to the center draining interior, smooth walls (no ledges) and the conical bottom. This product is manufactured in an ISO 9001:2015 Certified facility. Compares to Agilent Maximum Recovery vials. Use with 11mm aluminum crimp caps. These lower cost center draining vials have serum finish tops that provide the best seal for chromatography, when used properly with correct aluminum crimp caps.