Learn More
Microsolv Technology Corporation Vials, Crimp Top, Glass. Clear, 1.8ml, Max Recovery™. An 11mm Serum Finish with 12x32mm outer dimensions. For use as an autosampler vial or storage bottle. 100/PK.
Catalog No. 502270568
Encompass_Preferred
First hydrolytic, Class A, Type 33, Borosilicate Glass. Features a 12ul residual volume for most autosamplers due to the center draining interior and conical bottom. This product is manufactured in an ISO 9001:2015 Certified facility. Use with 11mm aluminum crimp caps. These lower cost center draining vials have serum finish tops that provide the best seal for chromatography, when used properly with correct aluminum crimp caps.
This item is not returnable. View return policy
Catalog No. 50-227-0568 Supplier Microsolv Technology Corporation Supplier No. 9512C00CV
First hydrolytic, Class A, Type 33, Borosilicate Glass. Features a 12ul residual volume for most autosamplers due to the center draining interior and conical bottom. This product is manufactured in an ISO 9001:2015 Certified facility. Use with 11mm aluminum crimp caps. These lower cost center draining vials have serum finish tops that provide the best seal for chromatography, when used properly with correct aluminum crimp caps.