Learn More
Microsolv Technology Corporation Vials, EPA Style, Glass. Amber, 20ml. 57x27.5mm outer dimensions. For use as an EPA test vial or storage bottle. ND24 threads. 100/PK.
Catalog No. 502271069
Encompass_Preferred
First hydrolytic, Class A, Type N51A, Borosilicate Glass, amberized. Amber vials are often used for light sensitive compounds and considered low actinic. Fill volume is approximately 23.3ml. Use 24mm screw caps that have 24-414 threads, sold separately. Lower cost for budget minded laboratories.
This item is not returnable. View return policy
Catalog No. 50-227-1069 Supplier Microsolv Technology Corporation Supplier No. 970210AV
First hydrolytic, Class A, Type N51A, Borosilicate Glass, amberized. Amber vials are often used for light sensitive compounds and considered low actinic. Fill volume is approximately 23.3ml. Use 24mm screw caps that have 24-414 threads, sold separately. Lower cost for budget minded laboratories.