Learn More
Microsolv Technology Corporation Vials, EPA Style, Glass. Clear, 30ml. 70x27.5mm outer dimensions. For use as an EPA test vial or storage bottle. ND24 threads. 100/PK.
Catalog No. 502271073
Encompass_Preferred
First hydrolytic, Class A, Type 33, Borosilicate Glass. Fill volume is approximately 31ml. Use 24mm screw caps that have 24-414 threads, sold separately. Lower cost for budget minded laboratories.
This item is not returnable. View return policy
Catalog No. 50-227-1073 Supplier Microsolv Technology Corporation Supplier No. 970310CV
First hydrolytic, Class A, Type 33, Borosilicate Glass. Fill volume is approximately 31ml. Use 24mm screw caps that have 24-414 threads, sold separately. Lower cost for budget minded laboratories.