Learn More
Electron Microscopy Sciences Vials for Cocoon Box
SDP
Catalog No. 5031086
Encompass_Preferred
Made from polystyrene and they are tough and crystal clear and come with a friction lid. Vial DB30 is designed for the Cocoon Box BE3D (64300-3D)
Catalog No. 50-310-86 Supplier Electron Microscopy Sciences Supplier No. 6430230
Made from polystyrene and they are tough and crystal clear and come with a friction lid. Vial DB30 is designed for the Cocoon Box BE3D (64300-3D)