Learn More
Electron Microscopy Sciences Vials for Cocoon Box
SDP
Supplier:  Electron Microscopy Sciences 6430220
Encompass_Preferred
Made from polystyrene and they are tough and crystal clear and come with a friction lid. Vial DB20 is designed for the cocoon Box BE3B (64300-3B)
Catalog No. 50-310-89