Learn More
Electron Microscopy Sciences Vials for Cocoon Box
SDP
Supplier:  Electron Microscopy Sciences 6430210
Encompass_Preferred
Made from polystyrene and they are tough and crystal clear and come with a friction lid. Vial DB10 is designed for the Cocoon Box BE4B (64300-4B)
Catalog No. 50-310-92