Learn More
Ace Glass, Inc. Vials, headspace, 10mL, flat bottom, 23x48mm, 20mm tapered finish top, pk/100
SDP
Catalog No. 50978408
Encompass
10mL clear borosilicate glass headspace vials. Vial is 23x48mm and features a flat bottom. Top is 20mm with a tapered finish. Meets USP type 1, class A specifications. 100 per pack.
Catalog No. 50-978-408 Supplier Ace Glass, Inc. Supplier No. 570728
10mL clear borosilicate glass headspace vials. Vial is 23x48mm and features a flat bottom. Top is 20mm with a tapered finish. Meets USP type 1, class A specifications. 100 per pack.