Learn More
Ace Glass, Inc. Vials, headspace, 20mL, flat bottom, 23x75mm, 20mm standard finish top, cs/1000
SDP
Catalog No. 50978406
Encompass
20mL clear borosilicate glass headspace vials. Vial is 23x75mm and features a flat bottom. Top is 20mm with a standard finish. Meets USP type 1, class A specifications. 1000 per case.
Catalog No. 50-978-406 Supplier Ace Glass, Inc. Supplier No. 570707
20mL clear borosilicate glass headspace vials. Vial is 23x75mm and features a flat bottom. Top is 20mm with a standard finish. Meets USP type 1, class A specifications. 1000 per case.