Learn More
Ace Glass, Inc. Vials, headspace, 20mL, flat bottom, 23x75mm, 20mm tapered finish top, pk/100
SDP
Catalog No. 509726
Encompass
Vials, headspace, 20mL, flat bottom, 23x75mm, 20mm tapered finish top, pk/100
Catalog No. 50-972-6 Supplier Ace Glass, Inc. Supplier No. 570730
Vials, headspace, 20mL, flat bottom, 23x75mm, 20mm tapered finish top, pk/100