Learn More
Ace Glass, Inc. Vials, headspace, 20mL, round bottom, 23x75mm, 20mm tapered finish top, pk/100
SDP
Catalog No. 50978409
Encompass
20mL clear borosilicate glass headspace vials. Vial is 23x75mm and features a round bottom. Top is 20mm with a tapered finish. Meets USP type 1, class A specifications. 100 per pack.
Catalog No. 50-978-409 Supplier Ace Glass, Inc. Supplier No. 570736
20mL clear borosilicate glass headspace vials. Vial is 23x75mm and features a round bottom. Top is 20mm with a tapered finish. Meets USP type 1, class A specifications. 100 per pack.