Learn More
Ace Glass, Inc. Vials, headspace, 6mL, flat bottom, 22x38mm, 20mm tapered finish top, cs/1000
SDP
Supplier:  Ace Glass, Inc. 570710
Encompass
Vials, headspace, 6mL, flat bottom, 22x38mm, 20mm tapered finish top, cs/1000
Catalog No. 50-972-0