Learn More
Microsolv Technology Corporation Vials, Headspace, Crimp Top, Glass. Clear. 10ml, 20mm Serum Finish, 20x54.5mm outer dimensions, Flat Bottom. Varian Compliant. 100/PK.
Catalog No. 502271221
Encompass_Preferred
First hydrolytic, Class A, Type N51A pharmaceutical grade, Borosilicate Glass. Use with aluminum or steel crimp caps for proper sealing. Compatible with Varian and other autosamplers.
This item is not returnable. View return policy
Catalog No. 50-227-1221 Supplier Microsolv Technology Corporation Supplier No. 95025VN10C
First hydrolytic, Class A, Type N51A pharmaceutical grade, Borosilicate Glass. Use with aluminum or steel crimp caps for proper sealing. Compatible with Varian and other autosamplers.