Learn More
Microsolv Technology Corporation Vials, Headspace, Crimp Top, Glass. Clear, 6ml, 20mm Serum Finish, 22x38mm outer dimensions, Flat Bottom with a Long Neck. Agilent Compatible. 100/PK.
Catalog No. 502270962
Encompass_Preferred
First hydrolytic, Class A, Borosilicate Glass, Use with aluminum or steel crimp caps for proper sealing. Compatible with Agilent, Carlo Erba, Dani and other Headspace autosamplers.
This item is not returnable. View return policy
Catalog No. 50-227-0962 Supplier Microsolv Technology Corporation Supplier No. 95025AG06C
First hydrolytic, Class A, Borosilicate Glass, Use with aluminum or steel crimp caps for proper sealing. Compatible with Agilent, Carlo Erba, Dani and other Headspace autosamplers.