Learn More
Microsolv Technology Corporation Vials, Headspace, Crimp Top, Glass. Clear. 20ml, 20mm Serum Finish, 22.5x75.5mm outer dimensions, Special Top and Flat Bottom for use with SPME Caps. CTC Compliant. 100/PK.
Catalog No. 502271220
Encompass_Preferred
First hydrolytic, Class A, Type N51A pharmaceutical grade, Borosilicate Glass. Use with aluminum or steel crimp caps with appropriate SPME Septa for proper sealing. Compatible with CTC Pal Solid Phase Micro-Extraction (SPME) autosamplers / Injectors.
This item is not returnable. View return policy
Catalog No. 50-227-1220 Supplier Microsolv Technology Corporation Supplier No. 95025SPME
First hydrolytic, Class A, Type N51A pharmaceutical grade, Borosilicate Glass. Use with aluminum or steel crimp caps with appropriate SPME Septa for proper sealing. Compatible with CTC Pal Solid Phase Micro-Extraction (SPME) autosamplers / Injectors.