Learn More
Microsolv Technology Corporation Vials, Headspace, Crimp Top, Glass. Amber, 20ml, 20mm Serum Finish, 22.5x75.5mm outer dimensions, Rounded Bottom. CTC Compliant. 100/PK.
Catalog No. 502271216
Encompass_Preferred
First hydrolytic, Class A, pharmaceutical grade, Borosilicate Glass, amberized and considered low actinic. Use with aluminum or steel crimp caps for proper sealing. Compatible with CTC Pal, Thermo, and other Headspace autosamplers. Not suitable with Thermo HS 250 or HS 500 autosamplers.
This item is not returnable. View return policy
Catalog No. 50-227-1216 Supplier Microsolv Technology Corporation Supplier No. 95025CT20A
First hydrolytic, Class A, pharmaceutical grade, Borosilicate Glass, amberized and considered low actinic. Use with aluminum or steel crimp caps for proper sealing. Compatible with CTC Pal, Thermo, and other Headspace autosamplers. Not suitable with Thermo HS 250 or HS 500 autosamplers.