Learn More
Microsolv Technology Corporation Vials, Headspace, Crimp Top, Glass. Amber. 10ml, 20mm Serum Finish, 23x46mm outer dimensions, Beveled Top and Rounded Bottom with a Tapered Neck. Perkin Elmer Compatible. 100/PK.
Catalog No. 502269911
Encompass_Preferred
First hydrolytic, Class A, Borosilicate Glass, amberized and considered low actinic. Use with aluminum or steel crimp caps for proper sealing. Compatible with Perkin Elmer, Tekmar and other Headspace autosamplers.
This item is not returnable. View return policy
Catalog No. 50-226-9911 Supplier Microsolv Technology Corporation Supplier No. 95025EPE10A
First hydrolytic, Class A, Borosilicate Glass, amberized and considered low actinic. Use with aluminum or steel crimp caps for proper sealing. Compatible with Perkin Elmer, Tekmar and other Headspace autosamplers.