Learn More
Microsolv Technology Corporation Vials, Headspace, Crimp Top, Glass. Amber, 10ml, 20mm Serum Finish, 22.5x46mm outer dimensions, Flat Bottom with a Long Neck. Agilent Compatible. 100/PK.
Catalog No. 502270160
Encompass_Preferred
First hydrolytic, Class A, Borosilicate Glass, amberized and considered low actinic. Use with aluminum or steel crimp caps for proper sealing. Compatible with Agilent and other Headspace autosamplers.
This item is not returnable. View return policy
Catalog No. 50-227-0160 Supplier Microsolv Technology Corporation Supplier No. 95025EAG10A
First hydrolytic, Class A, Borosilicate Glass, amberized and considered low actinic. Use with aluminum or steel crimp caps for proper sealing. Compatible with Agilent and other Headspace autosamplers.