Learn More
Microsolv Technology Corporation Vials, Headspace, Crimp Top, Glass. Amber. 6ml, 20mm Serum Finish, 22x38mm outer dimensions, Beveled Top and Flat Bottom with a Tapered Neck. Agilent Compatible. 100/PK.
Catalog No. 502269906
Encompass_Preferred
First hydrolytic, Class A, Borosilicate Glass, amberized and considered low actinic. Use with aluminum or steel crimp caps for proper sealing. Compatible with Agilent, Perkin Elmer and other Headspace autosamplers.
This item is not returnable. View return policy
Catalog No. 50-226-9906 Supplier Microsolv Technology Corporation Supplier No. 95025EAP06A
First hydrolytic, Class A, Borosilicate Glass, amberized and considered low actinic. Use with aluminum or steel crimp caps for proper sealing. Compatible with Agilent, Perkin Elmer and other Headspace autosamplers.