Learn More
Microsolv Technology Corporation Vials, Headspace, Crimp Top, Glass. Amber, 20ml, 20mm Serum Finish, 22.5x75.5mm outer dimensions, Rounded Bottom with a Tapered Neck. CTC Compliant. 100/PK.
Catalog No. 502270162
Encompass_Preferred
First hydrolytic, Class A, Borosilicate Glass, amberized and considered low actinic. Use with aluminum or steel crimp caps for proper sealing. Compatible with CTC Pal, and other Headspace autosamplers.
This item is not returnable. View return policy
Catalog No. 50-227-0162 Supplier Microsolv Technology Corporation Supplier No. 95025ECT20A
First hydrolytic, Class A, Borosilicate Glass, amberized and considered low actinic. Use with aluminum or steel crimp caps for proper sealing. Compatible with CTC Pal, and other Headspace autosamplers.